Learn More
Poltex Cryo Box Holder

Description
- Holds stacks of 5 x 5 x 2 in. cryo boxes.
- Pocket in front for pens, implements, tape.
- Durable PETG construction.
- Choose from wall mount (screws/anchors included), or magnet mount (powerful magnets included).
- Wall mount version either mounts to wall with screws/keyhold slots, or stands on counter (comes with non-slip rubber bumpons on base).
- Choose from 14 or 28 box capacity.
- See image for dimensions
Specifications
Specifications
| Color | Transparent |
| For Use With (Equipment) | Cryo Boxes |
| Holds | 28 Boxes |
| Material | Polycarbonate |
| Type | Cryogenic Box Storage Rack |
| Mounting Type | Magnet |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.