Learn More
United Scientific™ Cryo Coder For Star and Round Base Vials
SDPSCI_ED
Catalog No. S15895 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Blue
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15895 Blue
S15710 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15895 Supplier United Scientific™ Supplier No. P60114B
Add to Cart
Add to Cart
Easily identify different samples in your lab.
  • Designed to fit onto the top of our star and round base cryogenic vial closures.
  • Provide quick visual identification of specific vials for ease in organizing inventory.
  • Available in a range of colors.
  • Compatible with all United Scientific cryo vials except P60102 and P60103 skirted base cryo vials.
Specifications
Color Blue
Vial Type United Star and Round Base Cryo Vials
Product Type Cryo Coder