Learn More
Greiner Bio-One Cryo Rack for 48 Cryo Tubes, SBS Format, Low-profile Lid
Catalog No. 07001192 Shop All Greiner Bio-One Products
Change view
Click to view available options
Holds:
48 Cryotubes
Catalog No. Holds
07-001-192 48 Cryotubes
1 options
Catalog No. 07-001-192 Supplier Greiner Bio-One Supplier No. 803242
Add to Cart
Add to Cart
Organized, space‐efficient storage for cryogenic tubes designed to enhance freezer capacity and workflow efficiency for reliable daily use
  • Holds up to 48 Cryo tubes securely, enabling structured and high-density sample storage
  • Designed in SBS format, ensuring compatibility with automated systems, robotic handlers, and standardized storage equipment.
  • Manufactured from high‐quality Polypropylene, providing excellent durability, chemical resistance, and cold‐temperature stability for cryogenic applications.
  • Equipped with a low‐profile lid, allowing optimized freezer space usage and stackable storage without compromising accessibility
  • Supplied in a light blue color, supporting quick visual identification and streamlined sample organization across multiple racks or storage systems
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Shape Rectangular
Type Cryo Storage Box
Holds 48 Cryotubes