Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with External Thread Cap
GREENER_CHOICE

Catalog No. 07000259 Shop All Greiner Bio-One Products
Change view
Click to view available options
Closure Color:
Blue
Green
Natural
Red
Yellow
Style:
ID Writing Area
No Writing Area
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Style
07-000-259 Red ID Writing Area
07-000-257 Natural ID Writing Area
07-000-258 Natural No Writing Area
07-000-434 Green ID Writing Area
07-000-435 Yellow ID Writing Area
07-000-436 Blue ID Writing Area
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-000-259 Supplier Greiner Bio-One Supplier No. 126280
Add to Cart
Add to Cart
This item is not returnable. View return policy

Suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.

  • With Star foot base
  • Without ID writing area
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 150 white cap inserts
  • Polyethylener case
  • Dia. x Height: 12.4 x 47mm
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Red
Capacity (Metric) 2 mL
Bottom Shape Round (Starfoot)
Material Polypropylene
Thread Style External
Includes Starfoot Base, ID writing area and 150 white cap inserts
Length (Metric) 47 mm
Packaging Polyethylene case
Style ID Writing Area
Sterility Sterile
Closure Type Threaded
Diameter (Metric) 12.4 mm
Product Type Cryogenic Storage Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.