Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with Internal Thread Cap
GREENER_CHOICE

Catalog No. 07000253 Shop All Greiner Bio-One Products
Change view
Click to view available options
Closure Color:
Blue
Green
Natural
Orange
Red
Yellow
Bottom Shape:
Round
Round (Starfoot)
Style:
ID Writing Area
No Writig Area
No Writing Area
Thread Style:
External
Internal
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Bottom Shape Style Thread Style
07-000-253 Blue Round ID Writing Area External
07-000-270 Green Round (Starfoot) ID Writing Area Internal
07-000-267 Natural Round No Writing Area External
07-000-271 Natural Round (Starfoot) ID Writing Area Internal
07-000-255 Natural Round ID Writing Area Internal
07-000-274 Natural Round (Starfoot) No Writig Area External
07-000-256 Red Round ID Writing Area External
07-000-254 Green Round ID Writing Area External
07-000-432 Orange Round (Starfoot) ID Writing Area Internal
07-000-269 Blue Round (Starfoot) ID Writing Area Internal
07-000-272 Red Round (Starfoot) ID Writing Area Internal
07-000-431 Yellow Round (Starfoot) ID Writing Area Internal
07-000-273 Yellow Round (Starfoot) ID Writing Area External
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 07-000-253 Supplier Greiner Bio-One Supplier No. 121279
Add to Cart
Add to Cart

Made of high-grade polypropylene which guarantees high thermal resistance.

  • Internal thread has silicone gasket
  • With ID writing area
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 150 white cap inserts
  • Polyethylener case
  • With or without star foot base
  • Dia. x Height: 12
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens. Contents non-cytotoxic
Closure Color Blue
Autoclavable Not Autoclavable
Bottom Shape Round
Material Polypropylene
Thread Style External
Includes ID writing area and 150 white cap inserts
No. per Case 500
No. per Pack 100
Style ID Writing Area
Temperature Range (Metric) -196°C to +121°C
Volume (Metric) Working 1.8 to 2.0 mL
Sterility Sterile
Graduated Yes
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.