Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cryo.s Round Bottom 5 mL Polypropylene Tube with Internal Thread Cap
GREENER_CHOICE

Catalog No. 07000266 Shop All Greiner Bio-One Products
Change view
Click to view available options
Closure Color:
Blue
Green
Natural
Red
Yellow
Closure Type:
Screw Cap
Threaded
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Closure Type
07-000-266 Red Screw Cap
07-000-262 Natural Threaded
07-000-433 Yellow Threaded
07-000-264 Blue Screw Cap
07-000-265 Green Screw Cap
07-000-263 Natural Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-000-266 Supplier Greiner Bio-One Supplier No. 124273
Add to Cart
Add to Cart
This item is not returnable. View return policy

Made of high-grade polypropylene which guarantees high thermal resistance.

  • Internal thread has silicone gasket
  • With ID writing area
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 100 white cap inserts
  • Polyethylener case
  • Dia. x Height: 12.5 x 86mm
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens. Contents non-cytotoxic
Description With writing area
Closure Color Red
Autoclavable Not Autoclavable
Bottom Shape Round
Material Polypropylene
No. per Case 300
No. per Pack 50
Temperature Range (Metric) -196°C to +121°C
Volume (Metric) Working 4.5 to 5.0 mL
Sterility Sterile
Graduated Yes
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.