Learn More
Electron Microscopy Sciences CryoSleeve for Cryo Storage Canes
SDP
Supplier:  Electron Microscopy Sciences 6108210
Encompass_Preferred
Clear polyvinyl chloride. Ideal for enclosing Cryo Storage Canes for extra security during handling and storage. Fits over most other canes. Will not become brittle during freezing or thawing. Will not shred or tear after repeated use like cardboard.
Catalog No. 50-996-381
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.