Learn More
Cole-Parmer™ Homogenizer Cryo-block, Aluminum
Catalog No. 2665
Change view
Click to view available options
For Use With (Equipment):
96 Microcentrifuge or PCR Tubes
Catalog No. For Use With (Equipment)
26-65 96 Microcentrifuge or PCR Tubes
1 options
Catalog No. 26-65 Supplier Cole-Parmer™ Supplier No. 2665
Add to Cart
Add to Cart
Chilled Cryo-blocks are used for high-throughput cryogenic grinding and to preserve pesticides, RNA, proteins, and other temperature sensitive samples.
  • Enables high-throughput cryogenic grinding in the Geno/Grinder.
  • Maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis.
  • Preserves pesticides, RNA, and proteins for heat sensitive extraction methods.
  • Increases yields by grinding samples at sub-zero temperatures.
  • Durable aluminum construction.
  • Used two at a time in the Standard Clamp Assembly to keep it in balance.
Specifications
Product Type Cryo Block
Capacity (English) 0.02 oz.
Capacity (Metric) 0.6 mL
Description Each pair holds up to 192 Tubes for rapid and efficient sample preparation.
For Use With (Equipment) 96 Microcentrifuge or PCR Tubes
Holds 96 Vials
Material Aluminum
Packaging Quantity 2 Pk.