Learn More
StatLab CryoCare Bead Storage System
SDP
Catalog No. 22130503
Change view
Click to view available options
CryoCare Bead Storage System
Catalog No.
22-130-503 CryoCare Bead Storage System
1 options
Catalog No. 22-130-503 Supplier StatLab Supplier No. 11006
Store bacteria, fungi, yeasts, QC cultures or culture collections.
  • Mixed color 50 vials/pack - blue, yellow, green, white, red
  • Save money by stretching each culture to 15 or more cultures while meeting NCCLS standards
  • Can even store fastidious organisms such as Campylobacter, Neisseria, Haemophilus, and Gram negative anaerobes
  • Each vial contains ceramic beads in a broth base
  • To use, heavily inoculate the bead from a fresh, pure culture then freeze
  • Activate a single bead to yield a pure culture of active organism.
  • Store bacteria, fungi, yeasts, QC cultures or culture collections up to 10 years at -70°C or 1 to 3 years at -10°C.
Specifications
Color Assorted
Material Polypropylene
Shape Cylinder
Type Vial Storage Coder
Quantity 50/Pk.