Learn More
Eppendorf 3P Freezer Rack: CryoCube F740 series (3-compartment, MAX), 2 in/53 mm, side access, stainless steel
Supplier: Eppendorf 3P 6001071210
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.