Learn More
Eppendorf 3P CryoCube FC660h, 660 L, with LED interface, classic cooling liquids, and air-cooling, 208 V/60 Hz (US)
Supplier:  Eppendorf 3P F660420005
Encompass_Preferred
- Premium seals for optimal temperature uniformity- 2-stage cascade cooling system for reliable cooling processes- New ergonomic handle requires less force and provides easier access- Insulated inner lids help to maintain temperature stable when opening freezer- New high-efficiency fan, compressor, and condenser for lower energy consumption- Access ports for external alarm system for supervised sample storage- Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior- Compatible with CO2/LN2 injection system as backup for highest sample safety
Catalog No. NC1830098
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.