Learn More
Eppendorf 3P CryoCube F570n, 570 L, classic interface, green cooling liquids, air cooling, handle left side, 5 shelves, 115 V/60 Hz (US)
Supplier:  Eppendorf 3P F571200035
Encompass_Preferred
- Volume class: 570 L (20.1 ft3), e.g. up to 40,000 samples- Efficient insulation, composed of traditional insulation ("n"-version) and vacuum insulation panels ("h"-version), saves energy- Efficient compressor control system reduces cycle times to lower energy consumption and increases freezer longevity- Automatic vent port on front door of upright CryoCube models improves energy consumption and enables fast sample access- Lockable main switch to prevent switch-offs by accident- Location for chart recorder for data documentation- Ergonomic door handle of CryoCube models based on Eppendorf PhysioCare Concept requires less force and provides easier access- Air-cooled as well as water-cooled versions for further energy saving- Stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
Catalog No. NC1949621
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.