Learn More
R&D Systems™ CryoDefend-Cell Lines (5 x 10 mL)
SDP
Catalog No. CCM019 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
CCM019 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. CCM019 Supplier R&D Systems™ Supplier No. CCM019
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
5 x 10mL vials CryoDefend-Stem Cells Media
Media for defined, protein-free cryopreservation of cultured cells
Cryopreservation can be used to maintain cells at a low passage number and to reduce the risk of contamination by microbes and other cell types. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
Contains a defined serum substitute as well as an optimized concentration of a cryopreservative that increases the recovery and viability of healthy cells compared to conventional freezing media
This product contains
DMSO and is ready for use
Reduces experimental variability by using fully defined media
Suitable for all cell types
Specifications
Product Line R&D Systems
Quantity 1 Kit
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.