Learn More
GA International Cryo-DirectTAG™ - Cryogenic Labels for Direct Thermal Barcode Printers
Supplier: GA International DFP7C11WH
These cryogenic barcode labels are compatible with direct thermal printers. The thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microcentrifuge tubes, microtubes, microplates, glass tubes, freezer boxes, plastic bags, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage. These labels have a strong affinity for glass, polypropylene, polyethylene, and other plastic surfaces, and will retain their adhesion to tubes through multiple freeze-thaw cycles without cracking or peeling off. The direct thermal cryogenic labels are compatible with most cryogenic storage systems, without any hindrance or risk of being scraped off.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.