Learn More
Electron Microscopy Sciences Cryogenic Wrap Around Labels, 15/16" x 2.375"
SDP
Supplier:  Electron Microscopy Sciences 7702818
Encompass_Preferred
These labels are made from top quality materials and are designed specially for cryogenic use. Sub-zero use, features a 15/16" x 15/16" frosted area for thermal printing or marking with cryo markers. Rest of label is clear. Wraps the cryo-tube for a protective cover. Clear area for viewing contents.
Size: 15/16" x 2.375"
Use: Sub-zero use
Catalog No. 50-322-15
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.