Learn More
Cole-Parmer PolarSafe Cryogenic Storage Dewar, 2L, with 3 Round Canister Racks
Supplier:  Cole-Parmer 4426059
Encompass_Preferred
Facilitating cryogenic management, these PolarSafe storage dewars come with stainless steel racks to accommodate boxes or canisters (depending on model). With high thermal efficiency, each unit provides long term storage in liquid nitrogen for up to 200 days without refill, under normal operating conditions (may vary by model). The 95L and 115L models feature a lockable lid with temp and fluid level display. Enhance monitoring and ease of transportation with these Cole-Parmer Polarsafe Dewar Accessories which includes Smart Caps, trolleys, dewar monitor, and level rulers.
Catalog No. NC1865880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.