Learn More
Chemglass Life Sciences Cryogenic Vial, 1.2mL, External Thread, Self-Standing, Sterile, 50/Pk, 500/Box
SDP
Supplier:  Chemglass Life Sciences CGN476201A
Encompass_Preferred
Cryogenic vial with special screw cap designed without silicone ring to avoid the contamination risk. • USP grade VI Polypropylene • Autoclave at 121°C• Manufactured in clean room • Clear graduations and white marking area • Sterilized by E-Beam • Non-pyrogenic, DNase/RNase free Internal-Thread Cryogenic Vials • Used for storage in the gas phase of liquid nitrogen • Stand upright design for the bottom of the tube allows for self-standing • Optional color inserts available separately for caps
Catalog No. 50-189-7758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.