Learn More
TWD Scientific LLC CRYOKING 2" 100-WELL POLYCARB
SDP
Supplier:  TWD Scientific LLC 980213
Encompass_Preferred
CryoKING 2" 100-Well Polycarbonate Freezer Boxes Made of tough polycarbonate, chemically resistant to alcohols and mild organic solvents Can be repeatedly frozen and thawed Compatible with 0.5ml, 1.5ml, 2.0ml or 5.0ml microtubes
Catalog No. NC2206142