Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CryoMed™ with OPC UA, General Purpose

Catalog No. TSCM48SA
Change view
Click to view available options
Capacity (Metric):
17 L
34 L
48 L
Type:
Controlled-rate Freezer with Printer
Controlled-rate Freezer without Printer
Electrical Requirements:
120 V, 50-60 Hz
120V, 50-60Hz
220-230 V, 50-60 Hz
220-230V, 50-60Hz
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Type Electrical Requirements
TSCM48SA 48 L Controlled-rate Freezer without Printer 120 V, 50-60 Hz
TSCM34FA 34 L Controlled-rate Freezer with Printer 120V, 50-60Hz
TSCM34FV 34 L Controlled-rate Freezer with Printer 220-230V, 50-60Hz
TSCM34SA 34 L Controlled-rate Freezer with Printer 120V, 50-60Hz
TSCM34SV 34 L Controlled-rate Freezer with Printer 220-230 V, 50-60 Hz
TSCM48FA 48 L Controlled-rate Freezer with Printer 120V, 50-60Hz
TSCM48FV 48 L Controlled-rate Freezer with Printer 220-230V, 50-60Hz
TSCM48SV 48 L Controlled-rate Freezer without Printer 220-230V, 50-60Hz
TSCM17FV 17 L Controlled-rate Freezer with Printer 220-230V, 50-60Hz
TSCM17SV 17 L Controlled-rate Freezer without Printer 220-230V, 50-60Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSCM48SA Supplier Thermo Scientific™ Supplier No. TSCM48SA
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Thermo Scientific CryoMed Controlled-Rate Freezers with OPC UA are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • OPC UA (ethernet) serial communication protocol
  • Factory-issued certificates
  • Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • “Run last” feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Type Controlled-rate Freezer without Printer
Capacity (Metric) 48 L
Temperature Range -180°C to +50°C
Cabinet Material Stainless Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance cULus
Display Touchscreen user interface
Warranty Warranty varies by region. Contact your local sales representative for more information.
Sample Loading Front-loading
Dimensions (DxWxH) Exterior 24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior 13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data Outputs USB and OPC UA (ethernet)
On-Board Datalogging Yes
PC Programmability PC Interface software supported
Connectivity Thermo Fisher Connect compatible
Amperage 12 A
Plug Type NEMA 5-15 P
Electrical Requirements 120 V, 50-60 Hz
Vial Capacity <1185
Weight (English) 191 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.