Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Cryopan I, Serum-free Cell Freezing Medium, w/o: Human and Animal Components (100 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0792100
Encompass_Preferred
Cryopan I, Serum-free Freezing Medium, WITHOUT: Human and Animal Components. QUANTITY: 100 ml. MANUFACTURED BY: PAN-Biotech (Germany). Cryopan I consists of a chemically defined and optimized mixture of salts, sugar, 10 % DMSO and additional antifreeze-substances. Due to its serum-free formulation, Cryopan is especially suitable for the conservation of serum-free cultured cells. The optimized composition guarantees a high cell viability after the thawing process. Best results are attained for all cell types including primary human cells and human cell lines. As Cryopan I contains no animal or human components, a risk of contamination with viruses, BSE or disturbing proteins is excluded. STERILE liquid. STORAGE: at or below -20 degrees Celsius.
Catalog No. 50-238-8585
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.