Learn More
Thermo Scientific™ CryoPlus™ Series Racks and Holders
Catalog No. TF4000011
Change view
Click to view available options
Type:
Arrowhead Rack, Liquid Phase
Arrowhead Rack, Vapor Phase
Square Rack, Liquid Phase
Square Rack, Vapor Phase
Vertical Rack, Liquid Phase
Vertical Rack, Vapor Phase
Includes:
10 Cardboard Boxes (81 Cell Divider)
11 Cardboard Boxes (100 Cell Divider)
11 Cardboard Boxes (81 Cell Divider)
11 Stainless Steel Boxes (100 Cell Divider)
12 Cardboard Boxes (100 Cell Divider)
13 Cardboard Boxes (100 Cell Divider)
13 Cardboard Boxes (81 Cell Divider)
13 Stainless Steel Boxes (100 Cell Divider)
4 Boxes
5 Cardboard Boxes (100 Cell Divider)
6 Cardboard Boxes (100 Cell Divider)
7 Cardboard Boxes (100 Cell Divider)
8 Cardboard Boxes (100 Cell Divider)
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
TF4000011 Vertical Rack, Vapor Phase 4 Boxes
TF4000001 Square Rack, Vapor Phase 11 Cardboard Boxes (100 Cell Divider)
11-679-095 Square Rack, Vapor Phase 11 Stainless Steel Boxes (100 Cell Divider)
13-262-797 Square Rack, Vapor Phase 7 Cardboard Boxes (100 Cell Divider)
11-679-096 Square Rack, Vapor Phase 6 Cardboard Boxes (100 Cell Divider)
TF4000006 Square Rack, Liquid Phase 13 Cardboard Boxes (100 Cell Divider)
11-679-097 Square Rack, Liquid Phase 13 Stainless Steel Boxes (100 Cell Divider)
TF4000008 Square Rack, Liquid Phase 8 Cardboard Boxes (100 Cell Divider)
11-679-098 Square Rack, Liquid Phase 7 Cardboard Boxes (100 Cell Divider)
TF4000042 Square Rack, Liquid Phase 12 Cardboard Boxes (100 Cell Divider)
TF4000044 Arrowhead Rack, Liquid Phase 13 Cardboard Boxes (81 Cell Divider)
TF4000108 Arrowhead Rack, Liquid Phase 10 Cardboard Boxes (81 Cell Divider)
13-262-812 Arrowhead Rack, Vapor Phase 11 Cardboard Boxes (81 Cell Divider)
TF4000012 Vertical Rack, Liquid Phase 5 Cardboard Boxes (100 Cell Divider)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. TF4000011 Supplier Thermo Scientific™ Supplier No. 4000011
Add to Cart
Add to Cart
Increase the usability of Thermo Scientific™ CryoPlus™ LN2 storage systems with CryoPlus Series Racks and Holders.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
TRUSTED_SUSTAINABILITY
Specifications
Type Vertical Rack, Vapor Phase
For Use With (Equipment) CryoPlus 1,2,3,4
Includes 4 Boxes
Holds Vials
Rack Capacity 4 Boxes