Learn More
Cellular Engineering Technologies Cryopreservation Media (100mL)
Supplier:  Cellular Engineering Technologies MR1014
Encompass_Preferred
Media is highly versatile and can cryopreserve both adherent and suspension cells. Media has been tested with hematopoietic stem cells, mesenchymal stem cells, amniotic epithelial stem cells, cancer cells, endothelial cells, fibroblasts, and immune cells. Contains 10% DMSO.Media is shipped with dry ice.
Catalog No. NC2753087