Learn More
Cell Applications Inc Human Lung Microvascular Endothelial Cells Total Kit
SDP
Supplier:  Cell Applications Inc 540K05A
Encompass
Human Lung Microvascular Endothelial Cells Total Kit(540-05a) Cryopreserved (HLMVEC) 1 vial, adult(111-500) Microvascular Endothelial Cells Growth Media 500 ml(121-25-02) Extracellular Matrix Protein Coated T-25 Flasks (2 ea)(123-100) Attachment Factor Solution 100ml(090K) Subculture Reagent Kit
Catalog No. 50-195-9117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.