Learn More
BIOLIFE SOLUTIONS INC CryoStor® CS10 Freeze Media 10mL Vial
SDP
Supplier:  BIOLIFE SOLUTIONS INC 210373
Encompass
This item has a minimum qty of 4 per supplier requirements.
The CryoStor series of cell-specific, optimized freeze media, is designed to prepare and preserve cells in ultra low temperature environments (-70C to -196C). CryoStor CS10, pre-formulated with 10% DMSO, provides a safe, protective environment during the freezing, storage, and thawing process for cells and tissues with a requirement for 10% DMSO. Through modulating the molecular-biological response to the cryopreservation process, CryoStor provides for enhanced cell viability and functionality while eliminating the need for serum, proteins or high levels of cytotoxic agents.
Catalog No. NC0620116