Learn More
IMEB, Inc Specimen Object Disk, for use with Leica Cryostats, 30 mm
SDP
Catalog No. NC0427542
Encompass_Preferred
For use with Leica, Reichert, AO cryostats. Specimen object circular discs with outer O-ring for easy handling. 30mm diameter in circular embossed ring. For use on all frozen specimens. Not compatible with Leica CM1950 cryostats.
Catalog No. NC0427542 Supplier IMEB, Inc Supplier No. 370085
For use with Leica, Reichert, AO cryostats. Specimen object circular discs with outer O-ring for easy handling. 30mm diameter in circular embossed ring. For use on all frozen specimens. Not compatible with Leica CM1950 cryostats.