Learn More
BIOLIFE SOLUTIONS INC CryoStor CS5 Freeze Media 100mL Bottle
Supplier: BIOLIFE SOLUTIONS INC 205102
The CryoStor series of cell-specific, optimized freeze media, is designed to prepare and preserve cells in ultra low temperature environments (-70C to -196C). CryoStor CS5, pre-formulated with 5% DMSO, provides a safe, protective environment during the freezing, storage, and thawing process for cells and tissues with a requirement for 5% DMSO. Through modulating the molecular-biological response to the cryopreservation process, CryoStor provides for enhanced cell viability and functionality while eliminating the need for serum, proteins or high levels of cytotoxic agents.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.