Learn More
Simport™ Scientific Cryostore™ T314-481 Series Storage Boxes
Catalog No. 23045233 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-233 Blue
23-045-234 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-045-233 Supplier Simport™ Scientific Supplier No. T314481B
Add to Cart
Add to Cart
Cryostore™ T314-481 Series Storage Boxes for internally and externally threaded vials.
  • Made of polycarbonate
  • Transparent cover with numeric grid
  • Vented for drainage, stackable
  • Resistant to temperatures between +121°C to 196°C, but should only be used in the gas phase of liquid nitrogen
  • Autoclavable
Specifications
Color Blue
Dimensions (L x W x H) 133 x 133 x 81 mm
Holds 81 (3 to 4 mL Cryogenic Tubes)
Quantity 12/Cs.
Material Polycarbonate
Product Type Storage Box