Learn More
Andwin Scientific CRYOTAGS GR 43X19 1040/PK
SDP
Supplier:  Andwin Scientific 77565G
Encompass_Preferred
Suitable for laser printing, the 43X19 Cryo-Tags stay-flat labels in Green can be used on cryo-vials. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 1040 a pack.
Catalog No. 50-727-682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.