Learn More
Cole-Parmer Argos Technologies PolarSafe Sterile 2D Cryovials, 5 mL, Skirted-Bottom, External Thread; 50/Pk
Supplier:  Cole-Parmer 0439581
Encompass_Preferred
The PolarSafe cryogenic 2D vials are available in 2 and 5 mL sizes with internally or externally threaded caps. They are ideal for rapid processing of biological samples. The embedded Datamatrix code at the bottom of each vial, along with the corresponding Code 128 barcode on the body of the vial saves time and provides accurate organization, and tracking of valuable samples. The 2D vials can be safely stored in polycarbonate boxes that can be purchase separately. The storage boxes also features the 2D Datamatrix code and Code 128 barcode to provide automatic identification of both the box and stored vials.
Catalog No. 50-196-0646
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.