Learn More
Thomas Scientific Cap inserts for PolarSafe cryogenic vials, Blue
SDP
Supplier:  Thomas Scientific 1186P18
Encompass
Color code your samples Cap inserts are available in assorted colors and snap easily into place on both internally and externally threaded caps. Ideal for color coding and sample identification. Assorted colors sold 600 per pack (includes 100 of each color).
Catalog No. 50-146-1737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.