Learn More
Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials
Catalog No. 23045223 Shop All Simport Scientific Products
Change view
Click to view available options
Volume (Metric):
3 mL
4 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-045-223 4 mL
23-045-222 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-045-223 Supplier Simport™ Scientific Supplier No. T3094A
Add to Cart
Add to Cart
Designed for storing biological material, human or animal cells, at temperatures as low as -196°C. Cryovial™ External Thread with Lip Seal Cryogenic Vials should be used only in the gas phase of liquid nitrogen.
  • Made of specially formulated polypropylene
  • Cap features a long skirt and for one hand aseptic methods
  • Fast cap thread design for tightening or removal with a mere 1 1/4 turn and inside thread design will not contribute to contamination
  • Lip inside cap ensures leakproof seal even at low temperatures
  • Provided with white marking area for sample identification
  • Color coded by the use of a CAPINSERT™
  • Sterilized by gamma radiation
  • Packaged in unique tamperproof, resealable, safety-lock bags
Specifications
Color Natural
Material Polypropylene
Sterility Sterile
Volume (Metric) 4 mL
Thread Style External
Base Style Self-standing
Dnase Free Yes
Autoclavable Autoclavable
Certifications/Compliance CE; RNase, DNase, Pyrogen and DNA-free
For in vitro diagnostic (IVD) use only.