Learn More
Art Robbins Instruments CrysCam UV System, automated imaging system for protein crystals, imaging modes: visible, UV, fluorescent, optical magnification: 3.5x to 20x, camera resolution: 6.0 Mpixel, weight: 40 lbs, cold room compatible: true
SDP
Supplier:  Art Robbins Instruments 610900010
Encompass
CrysCam UV System, automated imaging system for protein crystals, imaging modes: visible, UV, fluorescent, optical magnification: 3.5x to 20x, camera resolution: 6.0 Mpixel, weight: 40 lbs, cold room compatible: true
Catalog No. NC3718675
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.