Learn More
Aremco Products Inc CRYSTAL BOND GLUE STICKS 5/PK
SDP
Supplier:  Aremco Products Inc 5091A
Encompass
Crystalbond 509 Glue Stick,5/pk,Provides excellent adhesion and minimizes clogging of diamond tools compared to waxes,Size- 7/8 x 7 in ,Weight: .2 lbs/stick ,Flow Point deg. F: 250,Viscosity, cps- 6,000 ,Color:Clear/Amber,Solvent 5
Catalog No. NC9668900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.