Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CrystalDrop™ Lids for CrystalQuick™ Microplates

Catalog No. 07000558 Shop All Greiner Bio-One Products
Change view
Click to view available options
Format:
Pre-greased
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
07-000-558 Pre-greased
07-000-562 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-558 Supplier Greiner Bio-One Supplier No. 609050

For use with CrystalQuick™ microplates. Greiner Bio-One CrystalDrop™ lids enables individual sealing of 96-wells.

  • 192-wells helps to place sample drops at predefined positions
  • Printed grid facilitates automated monitoring of crystallisation drops
  • Helps to conduct 288 sitting drop and 192 hanging drop experiments in parallel (total: 480 crystal
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Dimensions (L x W x H) 127.35 x 84.8 x 6.0 mm
Sterility Non-sterile
No. of Wells 96
No. per Case 40
Format Pre-greased
Plate Compatibility CrystalQuick Microplates

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.