Learn More
Mitegen In Situ 1 Crystallization Plate, For Direct X-ray Diffraction Analysis, Minimizes Sample Handling, 20 Plate Pack
SDP
Supplier:  Mitegen INSITU01CL20
Encompass_Preferred
The In Situ 1 Crystallization Plate is specifically designed to streamline the protein crystallization workflow, enabling direct X-ray diffraction analysis without the need for traditional crystal harvesting and mounting. This innovative plate format significantly minimizes sample handling, reducing the risk of crystal damage or contamination during sensitive experiments. It provides an optimized environment for crystal growth and subsequent direct observation, making it ideal for high-throughput screening and detailed structural biology studies. Manufactured from materials chosen for their optical clarity and minimal X-ray background interference, these plates ensure high-quality imaging and reliable diffraction data. Their design facilitates efficient sample visualization and alignment, enhancing experimental reproducibility. This is a pack of 20 plates.
Catalog No. 501940808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.