Learn More
DURAN™ DURAN™ Crystallizing Dish without Spout
GREENER_CHOICESDP
Catalog No. 01674623 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
2000 mL
Catalog No. Size
01-674-623 2000 mL
1 options
Catalog No. 01-674-623 Supplier DURAN™ Supplier No. 213135906
Add to Cart
Add to Cart
DURAN™ Crystallizing Dish without Spout is made from borosilicate glass 3.3 and complies with DIN 12337. Its flat, wide shape promotes rapid, even solvent evaporation, making it ideal for crystallization and daily laboratory workflows.
  • Complies with DIN 12337
  • Manufactured from borosilicate glass 3.3 for excellent chemical and thermal resistance
  • Flat and wide shape allows solvents to evaporate quickly and evenly
  • Large ground base provides stable support during use
  • Suitable for transferring substances and liquids into different states of aggregation
  • Supports various vessels, apparatus, and equipment used in crystallization and related laboratory processes
TRUSTED_SUSTAINABILITY
Specifications
Product Type Crystallization Dish
For Use With (Equipment) Hot Plate, Desiccators
Includes Without Spout
Size 2000 mL