Learn More
Hampton Research HR3-092G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609180), 4 ul square drop well, 1 drop well, LBR, hydrophobic, low profile - 20 plate case
SDP
Supplier:  Hampton Research HR3092G
Encompass
HR3-092G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609180), 4 ul square drop well, 1 drop well, LBR, hydrophobic, low profile - 20 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-025-7
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.