Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ CS720 Chemical Resistant Nitrile Gloves

Catalog No. 17467324 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-467-324 Large
17-467-322 Small
17-467-323 Medium
17-467-325 X-Large
17-467-326 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-467-324 Supplier SHOWA™ Supplier No. CS720L09
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical resistant nitrile, fully coated, blue, seamless knitted liner, wicks away moisture, 12 in. length, rough finish, small.

  • A double-dipped nitrile coating on the SHOWA CS720 chemical-resistant gloves keeps wearers safe from harmful substances.
  • A rough finish nitrile palm coating enables good grip performance, whilst the powdered liner wicks away moisture to prevent slipping and discomfort.
  • The durable CS720 gloves also prevent oil and dust from entering, ensuring optimum tactile precision.
  • Rough Grip
  • Double-coated
  • Nitrile Polyester Liner
  • Seamless knit
  • Increased dexterity
  • Oil-resistant Chemical resistant
  • Forearm Protection
  • Anti-slip Grip

Specifications

Applications Laboratory
Color Blue
Finger Thickness (English) 48 mil
Finger Thickness (Metric) 1.23 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 48 mil
Palm Thickness (Metric) 1.23 mm
Product Type Chemical-resistant Gloves
Size Large
Thickness (English) 48 mil
Thickness (Metric) 1.23 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.