Learn More
CSAFE LLC CSafe Global 8L, 120hr, 15°C to 25°C, Controlled-Room-Temperature (CRT) Parcel Shipper - ISTA 7D Qualified - Vacuum Insulation Panel (VIP) Technology. Perfect for long duration shipments of CRT products
Supplier:  CSAFE LLC A08C120000U
Encompass_Preferred
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance for the safe transport of vital life-science products such as Vaccines, Biologics, Cell Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a 8L payload, qualified to maintain a strict 15°C to 25°C controlled-room-temperature (CRT) condition for 120 hours against ISTA 7D’s worst-case challenge testing. CSafe utilizes proprietary Phase Change Material (PCM) for a simplistic, universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 35L solution is further protected by its double-wall outer corrugate, making this shipper ideal as a reusable system; while also being cost effective for one-way single-use shipments.
Catalog No. NC1866812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.