Learn More
CSAFE LLC CSafe Global 8L, 120hr Frozen Parcel Shipper - ISTA 7D Qualified - Vacuum Insulation Panel (VIP) Technology. Perfect for long duration shipments of Vaccines, Biologics and frozen therapies; including COVID-19 shipments
Supplier:  CSAFE LLC A08F120000U
Encompass_Preferred
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance and versality for the safe transport of vital life-science products such as Vaccines, Biologics, Cell & Gene Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a payload capacity of 8L and has been qualified to maintain frozen temperatures for 120 hours against ISTA 7D’s worst-case challenge testing, utilizing a universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 8L solution is further protected by its double-wall outer corrugate, making this solution ideal as a reusable system; while also being cost effective for one-way single-use shipments.
Catalog No. NC1866817
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.