Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSAG2 Monoclonal Antibody (OTI6C5), TrueMAB™, OriGene
SDP

Catalog No. 501683531
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3531 Supplier OriGene Supplier No. TA810621
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The expression of the cancer-testis antigen Taxol resistanceassociated gene-3 (TRAG-3) is associated with acquired paclitaxel (Taxol) resistance, and is expressed in a variety of cancers - e.g., breast cancer, leukemia, and melanoma. Overexpressed in taxolresistant breast cancer line MDA 435(TR) and the doxorubicinresistant multiple myelanoma lines 8226/Dox(40) and 8226/MDR (10)V. Weakly expressed in kidney. TRAG-3 is an attractive target for immunotherapy of cancer.

Specifications

Antigen CSAG2
Applications Western Blot
Classification Monoclonal
Clone OTI6C5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CSAG2
Gene Accession No. Q9Y5P2
Gene Alias CSAG2
Gene Symbols CSAG2
Host Species Mouse
Immunogen Full length human recombit protein of human CSAG2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102723547
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.