Learn More
CSB/ERCC6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301345A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a DNA-binding protein that is important in transcription-coupled excision repair. The encoded protein has ATP-stimulated ATPase activity, interacts with several transcription and excision repair proteins, and may promote complex formation at DNA repair sites. Mutations in this gene are associated with Cockayne syndrome type B and cerebrooculofacioskeletal syndrome 1. Naturally-occurring readthrough transcription occurs between this gene and the adjacent PGBD3 gene (GeneID:267004), and results in a fusion protein that shares sequence with the product of each individual gene. The readthrough locus is represented by GeneID:101243544. [provided by RefSeq, Mar 2013].Specifications
| CSB/ERCC6 | |
| Polyclonal | |
| Unconjugated | |
| ERCC6 | |
| excision repair cross-complementing rodent repair deficiency complementation group 6, ARMD5, CKN2, COFS, COFS1, RAD26, ATP-dependent helicase ERCC6, Cockayne syndrome B protein, Cockayne syndrome group B protein, DNA excision repair protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2074 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q03468 | |
| ERCC6 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.