Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSB/ERCC6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560149
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0149 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0149 Supplier Bethyl Laboratories, Inc Supplier No. A301347AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a DNA-binding protein that is important in transcription-coupled excision repair. The encoded protein has ATP-stimulated ATPase activity, interacts with several transcription and excision repair proteins, and may promote complex formation at DNA repair sites. Mutations in this gene are associated with Cockayne syndrome type B and cerebrooculofacioskeletal syndrome 1. Naturally-occurring readthrough transcription occurs between this gene and the adjacent PGBD3 gene (GeneID:267004), and results in a fusion protein that shares sequence with the product of each individual gene. The readthrough locus is represented by GeneID:101243544. [provided by RefSeq, Mar 2013].

Specifications

Antigen CSB/ERCC6
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ERCC6
Gene Accession No. Q03468
Gene Alias excision repair cross-complementing rodent repair deficiency complementation group 6, ARMD5, CKN2, COFS, COFS1, RAD26, ATP-dependent helicase ERCC6, Cockayne syndrome B protein, Cockayne syndrome group B protein, DNA excision repair protein
Gene Symbols ERCC6
Host Species Rabbit
Immunogen Between 1161 and 1211
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2074
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.