Learn More
Cytiva Centrasette cassette Supor PES membrane 0 1 m removal rating 0 5 m EFA suspended screen
Catalog No. 5000052810 Shop All Cytiva Products
Encompass
Centrasette cassette Supor PES membrane 0 1 m removal rating 0 5 m EFA suspended screen
Catalog No. 50-000-52810 Supplier Cytiva Supplier No. PSM10F07
Centrasette cassette Supor PES membrane 0 1 m removal rating 0 5 m EFA suspended screen