Learn More
Cytiva Centrasette cassette Supor PES membrane 0 45 m removal rating 2 5 m EFA medium screen
Catalog No. 5000052809 Shop All Cytiva Products
Encompass
Centrasette cassette Supor PES membrane 0 45 m removal rating 2 5 m EFA medium screen
Catalog No. 50-000-52809 Supplier Cytiva Supplier No. PSM45F26
Add to Cart
Add to Cart
Request a Quote
Centrasette cassette Supor PES membrane 0 45 m removal rating 2 5 m EFA medium screen