Learn More
Cytiva Centrasette cassette Supor PES membrane 0 65 m removal rating 0 5 m EFA suspended screen
Catalog No. 5000053048 Shop All Cytiva Products
Encompass
Centrasette cassette Supor PES membrane 0 65 m removal rating 0 5 m EFA suspended screen
Catalog No. 50-000-53048 Supplier Cytiva Supplier No. PSM65F07
Centrasette cassette Supor PES membrane 0 65 m removal rating 0 5 m EFA suspended screen