Learn More
Description
- Based on standard sandwich enzyme-linked immune-sorbent assay technology
- 96-well plates pre-coated with a monoclonal antibody from mouse specific for M-CSF
- Store at 4°C for 6 months, at -20°C for 12 months; avoid multiple freeze-thaw cycles
Specifications
Specifications
| Calibration Range | 62.5 to 4000pg/mL |
| Description | CSF1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CSF1. |
| Detection Method | Colorimetric |
| Product Type | CSF1 (Human) ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Tissue Homogenates, Urine |
| Target Species | Human |
| For Use With (Application) | Quantification |
| Gene ID (Entrez) | 1435 |
| Gene Symbol | Gene Alias: MCSF,MGC31930 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.