Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2RB Monoclonal Antibody (OTI2B10), TrueMAB™, OriGene
SDP

Catalog No. 501681818
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1818 Supplier OriGene Supplier No. TA807872
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSF2RB is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP). Diseases associated with CSF2RB include Surfactant Metabolism Dysfunction, Pulmonary, 5 and Hereditary Pulmonary Alveolar Proteinosis.

Specifications

Antigen CSF2RB
Applications Western Blot
Classification Monoclonal
Clone OTI2B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CSF2RB
Gene Accession No. P32927
Gene Alias CD131, CDw131, IL3RB, IL5RB, SMDP5, betaGMR
Gene Symbols CSF2RB
Host Species Mouse
Immunogen Full length human recombit protein of human CSF2RB produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.