Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF3 Rabbit anti-Human, Macaque, Unconjugated, Clone: CSF3/3166R, Abnova™

Catalog No. 89339496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-496 Supplier Abnova Corporation Supplier No. RAB00540
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CSF3 Monoclonal antibody specifically detects CSF3 in Human, Macaque samples. It is validated for IHC-P.

Specifications

Antigen CSF3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CSF3/3166R
Conjugate Unconjugated
Description colony stimulating factor 3 (granulocyte)
Gene CSF3
Gene Alias G-CSF, GCSF, MGC45931
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CSF3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Human, Macaque
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.