Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSP alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPG76 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPG76 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPG76 Supplier Invitrogen™ Supplier No. PA1776
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-776 detects Cysteine String Protein Alpha in mouse and rat brain samples. PA1-776 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~34 kDa protein representing Cysteine String Protein Alpha in rat and mouse brain homogenates. The PA1-776 immunogen is a synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP. This sequence is conserved across many species including mouse, human and bovine. This peptide (Cat. # PEP-287) is available for use in neutralization and control experiments.

Cysteine string protein alpha (CSPalpha) is a synaptic vesicle protein that contains a DNA-J domain characteristic of Hsp40 chaperones. Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain. These proteins play an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release. CSP proteins may act as presynaptic chaperones that assist in maintaining synaptic function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSP alpha
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Dnajc5
Gene Accession No. P60904, P60905, Q9H3Z4
Gene Alias 2610314I24Rik; AU018536; Ceroid-lipofuscinosis neuronal protein 4; CLN4; CLN4B; Csp; CSP alpha; cysteine string protein; cysteine string protein alpha; DnaJ (Hsp40) homolog, subfamily C, member 5; DnaJ heat shock protein family (Hsp40) member C5; dnaJ homolog subfamily C member 5; Dnajc5; DNAJC5A; mir-941-2; mir-941-3; mir-941-4; mir-941-5; NCL
Gene Symbols Dnajc5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13002, 79130, 80331
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.